Learn More
Medchemexpress LLC Gamma-glutamylphenylalanine | 7432-24-8 | MFCD00002629 | 99.8% | 294.30 g/mol | C14H18N2O5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10139925MG
Encompass_Preferred
γ-Glu-Phe (gamma-glutamylphenylalanine) is a gamma-linked dipeptide composed of glutamate and phenylalanine, provided as a high-purity research reagent and endogenous metabolite standard. It is used in biochemical and analytical studies, and is supplied with supporting documentation such as a certificate of analysis, safety data sheet, and product data sheet.
  • High purity suitable for analytical applications.
  • Solid form for easy handling and storage.
  • Available in small milligram pack sizes for method development.
  • CAS-registered chemical identifier for unambiguous identification.
  • Comes with COA and SDS for traceability and safety compliance.
  • Applicable as a metabolite standard and reagent in biochemical assays.
Catalog No. 50-002-13030