Learn More
Medchemexpress LLC Gamma-glutamylphenylalanine | 7432-24-8 | MFCD00002629 | 99.8% | 294.30 g/mol | C14H18N2O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY101399R50MG
Encompass_Preferred
Gamma-glutamylphenylalanine is an endogenous dipeptide composed of glutamate linked to phenylalanine. Supplied as a high-purity research standard, it is used in metabolite profiling, flavor chemistry, and analytical method development for chromatography and mass spectrometry. For research use only; not for human or clinical applications.
  • High purity suitable for analytical applications (99.8%).
  • Solid form, white to off-white.
  • Useful as a reference standard for metabolomics and chromatography.
  • Molecular weight 294.30 g/mol.
  • Packaged quantity 50 MG.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-11141