Learn More
Medchemexpress LLC Gamma-glutamylserine | 5875-35-4 | 98.4% | C8H14N2O6 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13811910MG
Encompass_Preferred
γ-Glutamylserine is a small-molecule amino acid derivative that functions as a calcium receptor activator. Provided as a white to off-white solid with formula C8H14N2O6 (MW 234.21), it is used in research on calcium receptor signaling and disease models such as Parkinson's disease, diabetes, and obesity.
  • High purity (98.4%).
  • White to off-white solid appearance.
  • Soluble in DMSO at 100 mg/mL; may require sonication.
  • Stable when stored sealed at -80°C (powder) for up to 2 years.
  • Suitable for in vitro research on calcium receptor activity and metabolic disease models.
Catalog No. 50-002-60451