Learn More
Sigma Aldrich Fine Chemicals Biosciences gamma Glutamyltransferase
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MAK0891KT
Encompass
I3-Glutamyltransferase (GGT EC 2.3.2.2) is a membrane-bound protein that catalyzes the transfer of -glutamyl moieties to acceptor molecules such as amino acids or peptides. GGT plays a key role in the»-glutamyl cycle a critical pathway for glutathione homeostasis as well as the detoxification of xenobiotics. Serum GGT levels can be elevated in many pathophysiological conditions such as cardiovascular disease chronic liver disease and metabolic syndrome. Elevated serum levels of GGT can also be indicative of oxidative stress.
Catalog No. 50-290-4324