Learn More
Medchemexpress LLC Gamma-secretase inhibitor I | 133407-83-7 | 97.6% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12023450MG
Encompass_Preferred
Z-LLNle-CHO (Z-Leu-Leu-Nle-CHO) is a peptide aldehyde gamma-secretase inhibitor used in biochemical and cell-based research. It blocks Akt-mediated pro-survival signaling and has been reported to induce caspase- and ROS-dependent apoptosis, making it useful for cancer research applications.
  • Gamma-secretase inhibitor for biochemical and cellular assays.
  • Induces caspase- and ROS-dependent apoptosis by blocking Akt signaling.
  • Used in cancer research models, including breast cancer and leukemia.
  • High purity suitable for research use (~97.6%).
  • Molecular formula C26H41N3O5; molecular weight 475.62 g/mol.
  • Supplied as a 50 MG pack for laboratory use.
Catalog No. 50-002-34436