Learn More
CROSSTEX INTERNATIONAL GAMMA STERILIZATION IND LABELS
Supplier: CROSSTEX INTERNATIONAL NMCPIR03
12 mm Printed Dot - Self-adhesive, nontoxic, chemical process indicators change from yellow to red upon exposure to gamma or electron beam radiation. Labels are coated with a UV blocking agent for protection from exposure to UV light. A minimum of 15kGy (1.5Mrad) irradiation is recommended for sufficient endpoint color development. At greater dosage levels, the radiation indicators exhibit a darker shade of red. Indicators are stable for at least 18 months from date of manufacture when they are stored at room temperature. 5000 Indicators per roll.