Learn More
Cayman Chemical γ-Tocotrienol, 14101-61-2, 1 mg
SDP
Supplier:  Cayman Chemical 100084941
Encompass_Preferred
Spontaneously hypertensive rats fed a diet which includes γ-tocotrienol show significant reduction of systolic blood pressure and an increase of nitric oxide synthase activity in blood vessels.{14293} γ-Tocotrienol at 20 µM induces apoptosis of malignant sympathoadrenal (+SA) murine mammary epithelial cells.{14294}
Catalog No. NC0104797