Learn More
Medchemexpress LLC Gancaonin M | 129145-51-3 | 1 MG

Supplier: Medchemexpress LLC HYN39221MG
Gancaonin M is a flavonoids product that can be isolated from the roots of Glycyrrhiza uralensis Fisch. It is intended for research purposes only.
- Purity of 97.13%
- Molecular weight of 352.38
- Chemical formula of C21H20O5
- Appearance as a solid
- Color is light yellow to yellow
- Classified as a flavonoid and isoflavone
- Isolated from plants, Leguminosae, Glycyrrhiza uralensis Fisch.
- Store at -20°C and protect from light
- When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.