Learn More
Selleck Chemical LLC Ganciclovir 250mg 82410-32-0 RS-21592, BW-759
SDP
Supplier:  Selleck Chemical LLC S1878250MG
Encompass_Preferred
Ganciclovir (RS-21592, BW-759) is an antiviral drug for feline herpesvirus type-1 with IC50 of 5.2 ?M in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1728