Learn More
Apexbio Technology LLC Ganciclovir, 82410-32-0, MFCD00870588, 50 mg
SDP
Supplier:  Apexbio Technology LLC B209750
Encompass_Preferred
MF: C9H13N5O4, MW: 255.23, Solubility: 6.95 mg/mL in DMSO with gentle warming, 25.6 mg/mL in H2O with gentle warming,insoluble in ETOH. Ganciclovir (DHPG) is a potent antiviral drug used to treatment of cytomegalovirus (CMV) infections.
Catalog No. 50-101-1678