Learn More
Medchemexpress LLC Ganciclovir | 82410-32-0 | MFCD00870588 | >99.89% | 255.23 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1363710MM/1ML
Encompass_Preferred
Ganciclovir (BW 759) is a nucleoside analogue and an orally active antiviral agent. It is known for its activity against Cytomegalovirus (CMV), as well as other members of the herpes group and various DNA viruses. Ganciclovir effectively inhibits the in vitro replication of human herpes viruses (HSV 1 and 2, CMV) and adenovirus serotypes 1, 2, 4, 6, 8, 10, 19, 22 and 28. It also demonstrates an IC50 of 5.2 μM for feline herpesvirus type-1 (FHV-1) and has the ability to diffuse into the brain.
  • Acts as an orally active antiviral agent
  • Effective against Cytomegalovirus (CMV)
  • Inhibits replication of human herpes viruses and adenoviruses
  • Demonstrates activity against feline herpesvirus type-1
  • Can diffuse into the brain
Catalog No. 50-003-49133