Learn More
Medchemexpress LLC Ganciclovir mono-O-acetate | 88110-89-8 | 98.0% | 297.27 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY4134410MM/1ML
Encompass_Preferred
Ganciclovir mono-O-acetate is a derivative of Ganciclovir, which is a nucleoside analogue. This compound functions as an orally active antiviral agent, specifically demonstrating activity against Cytomegalovirus (CMV).
  • Derivative of Ganciclovir
  • Orally active antiviral agent
  • Active against CMV
Catalog No. 50-004-04537