Learn More
Medchemexpress LLC Ganciclovir mono-o-acetate | 88110-89-8 | 98.0% | 297.27 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY4134450MG
Encompass_Preferred
Ganciclovir mono-O-acetate is a derivative of Ganciclovir, an orally active antiviral nucleoside analogue effective against CMV. This product is a mono-O-acetate ester of Ganciclovir.
  • Derivative of Ganciclovir
  • Orally active antiviral nucleoside analogue
  • Effective against CMV
  • Available as a solid
  • High purity
Catalog No. 50-004-03543