Learn More
Medchemexpress LLC Ganciclovir sodium | 107910-75-8 | 99.92% | 277.21 | 1 G
SDP
Supplier:  Medchemexpress LLC HY13637A1G
Encompass_Preferred
Ganciclovir (BW 759) sodium is a nucleoside analogue and an orally active antiviral agent effective against CMV. It also exhibits in vitro activity against members of the herpes group and other DNA viruses. It inhibits the in vitro replication of human herpes viruses (HSV 1 and 2, CMV) and adenovirus serotypes 1, 2, 4, 6, 8, 10, 19, 22 and 28. Ganciclovir sodium has an IC50 of 5.2 μM for feline herpesvirus type-1 (FHV-1) and can diffuse into the brain.
  • Orally active antiviral agent
  • Effective against CMV
  • Inhibits in vitro replication of human herpes viruses (HSV 1 and 2, CMV)
  • Inhibits adenovirus serotypes
  • IC50 of 5.2 μM for feline herpesvirus type-1 (FHV-1)
  • Can diffuse into the brain
Catalog No. 50-003-47947