Learn More
Apexbio Technology LLC Ganciclovir sodium 107910-75-8 250mg
SDP
Supplier:  Apexbio Technology LLC C6499250
Encompass_Preferred
Ganciclovir sodium is a nucleoside analog antiviral agent that functions primarily by inhibiting viral DNA polymerase thus blocking viral DNA replication It exerts inhibitory activities in vitro against various DNA viruses including human herpesviruses (e g HSV) and adenovirus serotypes Experimental data indicate inhibitory activity against feline herpesvirus type 1 with an IC50 value of approximately 5 2 M Additionally Ganciclovir sodium shows the capacity to cross the blood-brain barrier Common research applications include antiviral pharmacology studies virology-focused experimental assays and investigations of viral etiology
Catalog No. 50-000-64209