Learn More
Selleck Chemical LLC Ganciclovir sodium 25mg 107910-75-8 RS-21592 sodium, Cytovene IV sodium
SDP
Supplier:  Selleck Chemical LLC S506525MG
Encompass_Preferred
Ganciclovir Sodium (RS-21592 sodium, Cytovene IV sodium) is the sodium salt form of ganciclovir, a synthetic, antiviral, purine nucleoside analog with antiviral activity, especially against cytomegalovirus (CMV). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8313