Learn More
TARGETMOL CHEMICALS INC GANDOTINIB 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T263825MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LY2784544(Gandotinib (LY2784544)) is a potent JAK2 inhibitor (IC50 3 nM) effective in JAK2V617F(Ki 0.245 nM). The selectivity is higher 8- and 20-fold than JAK1 and JAK3. purity: 99%
Catalog No. 50-258-0700