Learn More
Selleck Chemical LLC Ganetespib (STA-9090) 5mg 888216-25-9
SDP
Supplier:  Selleck Chemical LLC S11595MG
Encompass_Preferred
Ganetespib (STA-9090) is an HSP90 inhibitor with IC50 of 4 nM in OSA 8 cells, induces apoptosis of OSA cells while normal osteoblasts are not affected; active metabolite of STA-1474. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0968