Learn More
Medchemexpress LLC Ganfeborole hydrochloride | 2131798-13-3 | MFCD31657361 | 100.0% | 293.94 g/mol | C10H14BCl2NO4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10777550MG
Encompass_Preferred
Ganfeborole hydrochloride is the hydrochloride salt of a boron-containing small molecule inhibitor of Mycobacterium tuberculosis leucyl-tRNA synthetase (LeuRS). It is supplied for research use in antitubercular drug discovery and mechanistic studies, with reported biochemical potency (IC50) of 0.2 μM against Mtb LeuRS.
  • Potent LeuRS inhibitor with reported IC50 0.2 μM.
  • Boron-containing benzoxaborole scaffold provides a unique mechanism of action.
  • Hydrochloride salt form improves solubility for biochemical assays.
  • Suitable for biochemical and cellular studies of tuberculosis targets.
  • Available in small research quantities for early-stage studies.
Catalog No. 50-002-42749