Learn More
Medchemexpress LLC Ganoderenic acid E | 110241-23-1 | 5 MG

Supplier: Medchemexpress LLC HYN68675MG
Ganoderenic acid E is a lanostane-type triterpene, initially sourced from Plants Polyporaceae. It is intended for research use only and is not sold to patients. This product is also identified as a controlled substance and may not be available for sale in certain territories.
- Lanostane-type triterpene
- Sourced from plants, Polyporaceae
- Purity of 95.0%
- Solid appearance
- White to off-white color
- Classified as terpenoid and triterpene
- Store at 4°C, sealed, away from moisture
- If in solvent, store at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.