Learn More
Selleck Chemical LLC Ganoderic acid A S4753-5mg
SDP
Supplier:  Selleck Chemical LLC S47535MG
Encompass_Preferred
Ganoderic Acid A (GAA) is an active triangle compound in Ganoderma Lucidum It can inhibit the JAK-STAT3 signal pathway and inhibit cell proliferation vitality and ROS Ganoderic Acid A has analgesic antioxidant cytotoxicity protecting liver and anti -tumor activity
Catalog No. 50-310-9370