Learn More
Medchemexpress LLC Ganoderic acid A | 81907-62-2 | 516.67 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN1447
Encompass_Preferred
Ganoderic acid A inhibits the JAK-STAT3 signaling pathway and demonstrates the ability to suppress proliferation, viability, and reactive oxygen species (ROS). It can also enhance HLA class II-mediated antigen presentation and CD4+ T cell recognition of lymphoma at lower doses. It also promotes cisplatin-induced cell death.
  • Inhibits JAK-STAT3 signaling pathway.
  • Suppresses proliferation and viability.
  • Reduces reactive oxygen species (ROS) and DPPH.
  • Enhances HLA class II-mediated antigen presentation.
  • Promotes cisplatin-induced cell death.
  • Induces cell apoptosis in osteosarcoma cells.
  • Prolongs survival in EL4 challenged mice and decreases tumor metastasis.
Catalog No. 50-003-89883