Learn More
Medchemexpress LLC Ganoderic acid B | 81907-61-1 | MFCD09752803 | 99.6% | 516.67 g/mol | C30H44O7 | 10 MG

Supplier: Medchemexpress LLC HYN200610MG
Ganoderic acid B is a triterpene natural product isolated from the mushroom Ganoderma lucidum and supplied for research use in a 10 mg vial. The material is characterized with standard analytical documentation and is intended for in vitro biochemical assays and related research applications.
- Triterpene isolated from Ganoderma lucidum.
- High reported purity of 99.6%.
- Provided in a 10 mg research vial.
- Characterized with COA, HNMR, CNMR, RP-HPLC, and MS documentation.
- Soluble in DMSO at 50 mg/mL with ultrasonic assistance.
- Store solid at 4°C protected from light; in solution store at -80°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.