Learn More
Medchemexpress LLC Ganoderic acid C6 | 105742-76-5 | MFCD21333706 | 99.4% | 530.65 g/mol | C30H42O8 | 10 MG

Supplier: Medchemexpress LLC HYN246110MG
Ganoderic acid C6 is a natural triterpenoid with reported aldose reductase inhibitory activity. It is supplied as a high-purity research reagent intended for biochemical and disease-model studies and is not for human or clinical use.
- Aldose reductase inhibitory activity.
- High purity, reported 99.4%.
- Molecular formula C30H42O8; molecular weight 530.65 g/mol.
- Provided as a 10 MG solid for research applications.
- Intended for biochemical assays and disease-research models.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.