Learn More
Medchemexpress LLC Ganoderic acid D | 108340-60-9 | 514.65 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN151110MM/1ML
Encompass_Preferred
Ganoderic acid D is a highly oxygenated tetracyclic triterpenoid and a major active component of *Ganoderma lucidum*. It upregulates SIRT3 protein expression, inducing deacetylated cyclophilin D (CypD) by SIRT3. This compound inhibits energy reprogramming in colon cancer cells, including glucose uptake, lactate production, pyruvate, and acetyl-coenzyme production. Ganoderic acid D also induces apoptosis in HeLa human cervical carcinoma cells and inhibits the growth of numerous cancer cell lines.
  • Upregulates SIRT3 protein expression and induces deacetylated cyclophilin D (CypD).
  • Inhibits energy reprogramming in colon cancer cells.
  • Induces apoptosis in human cervical carcinoma cells.
  • Inhibits the growth of various cancer cell lines.
  • Exhibits anti-inflammatory activity.
  • Induces G2/M phase arrest.
Catalog No. 50-004-16417