Learn More
Medchemexpress LLC Ganoderic acid E | 98665-14-6 | 512.63 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN151210MM/1ML
Encompass_Preferred
Ganoderic acid E is a triterpenoid found in Ganoderma lucidum. It is for research use only and not for sale to patients.
  • It is a triterpenoid.
  • It is found in Ganoderma lucidum.
  • It exhibits cytotoxicity against human HeLa cells (IC50 101 μM), human HepG2 cells (IC50 1.44 x 10-4 μM), human HepG2(2.2.15) cells (IC50 1.05 x 10-4 μM), and mouse P388 cells (IC50 5.012 μM).
  • It inhibits 12-O-tetradecanoylphorbol-13-acetate-induced EBV-early antigen activation in human Raji cells (281 molar ratio).
  • Available in solid form (white to off-white).
  • Purity: 98.08%.
Catalog No. 50-004-14980