Learn More
Medchemexpress LLC Ganoderic acid H | 98665-19-1 | 572.69 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN15131MG
Encompass_Preferred
Ganoderic acid H is a lanostane-type triterpene isolated from *Ganoderma lucidum*. It has been shown to suppress the growth and invasive behavior of breast cancer cells by inhibiting the activity of transcription factors AP-1 and NF-kappaB signaling pathways.
  • Isolated from *Ganoderma lucidum*.
  • Suppresses growth of breast cancer cells.
  • Inhibits invasive behavior of breast cancer cells.
  • Modulates AP-1 signaling.
  • Modulates NF-kappaB signaling.
  • For research use only.
Catalog No. 50-004-15138