Learn More
Medchemexpress LLC GANT 61 | 500579-04-4 | 99.80% | 429.60 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY13901
Encompass_Preferred
GANT 61 is a potent inhibitor of Gli1 and Gli2, effectively targeting the Hedgehog/GLI signaling pathway. This compound has demonstrated significant biological activity both in vitro and in vivo, impacting cell proliferation, viability, and tumor growth across various cancer models.
  • Inhibits Gli1 and Gli2 activity
  • Targets the Hedgehog/GLI pathway
  • Reduces clonogenic survival in human colon carcinoma cells
  • Induces apoptosis and inhibits cell viability in pancreatic cancer stem cells
  • Inhibits motility, invasion, and migration of cancer stem cells
  • Decreases tumor growth in xenograft models
  • Enhances the effects of chemotherapeutic drugs for neuroblastoma
Catalog No. 50-003-48671