Learn More
Cayman Chemical Gap 26trIfluoROCTAte salt 10mg
SDP
Supplier:  Cayman Chemical 3662510 MG
Encompass_Preferred
A connexin-mimetic peptide; inhibits phenylephrine-induced rhythmic contractile activity in isolated rabbit superior mesenteric arterial strips (IC50 = 28.4 µM); inhibits photoliberated Ins(1,4,5)-P3-induced ATP release in ECV304 cells at 0.25 mg/ml; decreases the infarct size in a rat model of myocardial infarction induced by left coronary artery occlusion at 0.5, 1, and 10 UG/kg
Catalog No. 50-291-3793