Learn More
LICOR GAPDH Rabbit Monoclonal Antibody for Normalization (100 uL)
Supplier:  LICOR 92642216
Encompass_Preferred
Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) is a constitutively expressed housekeeping protein (HKP). The GAPDH primary antibody can be used as an internal loading control for normalization. The expression of GAPDH, or any HKP, should be validated to ensure that its expression does not change under experimental conditions. Once validated, GAPDH primary antibodies can be used for the detection of GAPDH when performing multiplex Western blot detection. Detect GAPDH Rabbit Monoclonal Antibody with IRDye Goat anti-Rabbit or IRDye Donkey anti-Rabbit secondary antibodies.
Catalog No. NC2036053