Learn More
Selleck Chemical LLC Garcinia cambogia extract E2863-25MG
SDP
Supplier:  Selleck Chemical LLC E286325MG
Encompass_Preferred
Garcinia cambogia extract has hypocholesterolemic effect and used for obesity treatment (-)-hydroxycitrate (HCA) the chief compound of standardized Garcinia cambogia extract is a competitive inhibitor of ATP-citrate-lyase
Catalog No. 50-000-40518