Learn More
Cayman Chemical Garcinol, 78824-30-3, 10 mg
SDP
Supplier:  Cayman Chemical 1056610
Encompass_Preferred
An inhibitor of the HATs p300 and pCAF (IC50 = 7 and 5 μM, respectively) that also inhibits the HAT Gcn5; promotes neurogenesis and ex vivo expansion of human hematopoietic stem cells; induces apoptosis in several types of cancer cells and has anti-inflammatory actions
Catalog No. NC0393469
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.