Learn More
STEMCELL Technologies Garcinol, Size: 5 mg
SDP
Supplier:  STEMCELL Technologies 72452
Encompass
Garcinol is an inhibitor of histone acetyltransferases (HATs) p300 and pCAF (IC₅₀ 7 and 5 respectively) (Balasubramanyam et al.). It also inhibits the HAT GCN5 in Cryptococcus neoformans inducing temperature sensitivity and impairing growth (O’Meara et al.).
Catalog No. 50-239-0376