Learn More
Abcam Garcinol, HAT p300 and PCAF inhibitor, 5MG
Supplier:  Abcam AB1415035MG
Encompass_Preferred
MW 602.8 Da, Purity >95%. Potent histone acetyltransferase p300 and PCAF inhibitor (IC₅₀ values are 7 and 5 μM for p300 and PCAF respectively). Pleiotropic agent. Strongly inhibits chromatin transcription and induces apoptosis. Inhibits histone acetylation in vivo. Antioxidant and chemoprotective agent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6558