Learn More
Cayman Chemical Garcnon D 10mg
SDP
Supplier:  Cayman Chemical 3090610 MG
Encompass_Preferred
A xanthone with diverse biological activities; increases proliferation of C17.2 NPCs at 5 µM; inhibits aggregation of Aβ42 in vitro at 3 µM and inhibits BACE1 by 62.7% at 100 µM; scavenges peroxynitrite radicals (IC50 = 26.4 µM)
Catalog No. 50-298-9926