Learn More
Medchemexpress LLC HY-103697A 50mg , Gardiquimod (diTFA) CAS:1159840-61-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY103697A50MG
Encompass_Preferred
HY-103697A 50mg Gardiquimod (diTFA) CAS: 1159840-61-5 Gardiquimod diTFA, an imidazoquinoline analog, is a TLR7/8 agonist. Gardiquimod diTFA could inhibit HIV-1 infection of macrophages and activated peripheral blood mononuclear cells (PBMCs). Gardiquimod diTFA specifically activates TLR7 when used at concentrations below 10 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1259