Learn More
Medchemexpress LLC GARS Human sf9 Hi 10ug
SDP
Supplier:  Medchemexpress LLC HYP7412810UG
Encompass_Preferred
The GARS protein performs ATP-dependent ligation of glycine to its tRNA creating a Gly-AMP intermediate It also produces diadenosine tetraphosphate (Ap4A) by condensing two ATP molecules which is important for cellular regulation GARS may have a specific role in maintaining Ap4A homeostasis GARS Protein Human (sf9 His) is the recombinant human-derived GARS protein expressed by Sf9 insect cells with N-His labeled tag
Catalog No. 50-001-79068