Learn More
MSC Value Collection Pressure Washer, 4.0 GPM, Cold Water, Gas, AR Triplex Ceramic Pump
Supplier:  MSC 48352645
Encompass
This Value Collection pressure washer is designed for powerful cleaning tasks, featuring a powerful engine, a 4.0 GPM flow rate, and an AR Triplex ceramic plunger pump. It is engineered for cold water applications and includes a 50 ft hose for extended reach.
  • Equipped with an AR Triplex ceramic plunger pump.
  • Includes a 50 ft hose.
  • Features a 2-wheel cart frame for portability.
  • Operates with cold water.
  • Powered by gas.
  • Displacement of 420.0 cc.
  • Delivers a flow rate of 4.0 GPM.
  • Features a powerful engine.
  • Provides 3700 pressure.
  • Comes with a 3/8 in hose size.
Catalog No. 50-283-8970
Add to Cart
Request a Quote