Learn More
MSC Value Collection Pressure Washer, 5.0 GPM, 4000 PSI, Gas, Cold Water
Supplier:  MSC 48352710
Encompass
This pressure washer is designed for demanding cleaning tasks, featuring a gas-powered engine and delivering cold water at high pressure. It is equipped with an AR Triplex ceramic plunger pump and a Vanguard engine, ensuring reliable performance and durability.
  • Cold water type for versatile cleaning applications
  • Gas power for robust performance without electrical limitations
  • High flow rate of 5.0 GPM for efficient cleaning
  • Powerful 4000 PSI pressure to tackle tough dirt and grime
  • AR Triplex ceramic plunger pump for enhanced longevity and consistent output
  • 50.00 feet hose length provides extended reach
  • Integrated 2 wheel roll cage frame for easy portability and protection
Catalog No. 50-283-8973
Add to Cart
Request a Quote