Learn More
Chemglass Life Sciences Gas Dispersion Tube, Large Capacity, Fritted, Coarse Porosity
SDP
Supplier:  Chemglass Life Sciences CG22001
Encompass_Preferred
Gas Dispersion Tube, Large Capacity, Fritted, Coarse Porosity. Large capacity gas dispersion tube having a frit 25mm O.D. x 70mm long. Stem is 16mm O.D. x 200 mm above the frit. Overall length is approximately 280mm.
Catalog No. CG22001