Learn More
Perkin Elmer US LLC Gas/Liquid Separator for Flow Injection
Supplier:  Perkin Elmer US LLC B0507959
Encompass_Preferred
Glass Gas/Liquid Separater (B0193772) available separately. Note: Not for use with ICP-MS. Use Glass Gas/Liquid Separater (B0193772).
Catalog No. 50-904-2498