Learn More
Glass Expansion Gasket for PerkinElmer NexION 300/350/1000/1100/2000/2200/5000 ICP-MS Sampler Cone (W1040148)
SDP
Supplier:  Glass Expansion PE5111
Encompass_Preferred
Gasket for Sampler Cone, PKT 5 (W1040148). For PerkinElmer NexION 1000/2000/5000/300/350 ICP-MS. Use a new gasket each time the cone is re-installed, even if it is the same cone.
Catalog No. 50-190-7691