Learn More
Helix Medical Inc GASKET SILICONE CLAMP 1.5 PK25
Supplier:  Helix Medical Inc 7040015
Encompass_Preferred
Biomedical/pharmaceutical grade, platinum-cured silicone clamp gaskets promote crevice-free and protrusion-free processing. The gaskets offer exceptional biocompatibility and contain no contaminants or leachables. May be sterilized by autoclave, ethylene oxide, or gamma radiation.
Catalog No. NC1329721