Learn More
Fritsch Milling And Sizing Inc Gassing Lid Cpl. Zirconium Oxide P-5 Pl 150-500Ml
SDP
Supplier:  Fritsch Milling And Sizing Inc 50.6497.00
Encompass
Gassing lid P-5 pl with valves and seal ring made of zirconium oxide with steel casing
For all grinding bowls premium line 500 ml, 250 ml, 150 ml volume made of agate for PULVERISETTE 5 premium line.
By using this special lid, the grinding bowl can be used for grinding in inert gas and for mechanical alloying - quickly and safely. Gassing lids with Swagelok valves are available on request.
Catalog No. 50-153-4583
Only null left
Add to Cart
Request a Quote