Learn More
Medchemexpress LLC Gastric lipase protein, human (HEK293, His) | >95.0% | 500UG

Supplier: Medchemexpress LLC HYP70262A500UG
Recombinant human gastric lipase is supplied as a purified protein produced in HEK293 cells with a C-terminal His tag for affinity purification. It is provided as a lyophilized powder with reported purity and endotoxin data and is intended for biochemical and enzymatic research. Enzyme activity testing is ongoing and activity is not guaranteed.
- Expressed in HEK293 cells.
- C-terminal 6xHis tag for affinity purification.
- Purity: >95.0% by reducing SDS-PAGE.
- Endotoxin: <1 EU/μg (LAL).
- Predicted molecular mass 44.2 kDa; observed ~48-50 kDa due to glycosylation.
- Supplied as lyophilized powder; reconstitute according to datasheet and aliquot to avoid freeze-thaw.
- Storage: store at -20°C; after reconstitution stable at 4°C for one week or frozen for longer storage.
- Enzyme activity testing in progress; activity is not guaranteed.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.