Learn More
Medchemexpress LLC Gastrin-71 Mouse H 2ug | 2ug
SDP
Supplier:  Medchemexpress LLC HYP787032UG
Encompass_Preferred
Gastrin-71 protein acts as a potent stimulator of various physiological processes It activates the gastric mucosa promotes the production and secretion of hydrochloric acid and induces the release of digestive enzymes from the pancreas Gastrin-71 Protein Mouse (HEK293 Fc) is the recombinant mouse-derived Gastrin-71 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-94325