Learn More
Enzo Life Sciences Gastrin I (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900026
Encompass_Preferred
The Gastrin I (human), EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. Ready-to-use liquid color-coded reagents are provided to reduce error in the lab. Sensitivity: 7.27 pg/ml (range 39.1 - 10,000 pg/ml). Assay Time: 5 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human. Contents: GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-658-110