Learn More
Medchemexpress LLC Gastrin I, rat | 81123-06-0 | 99.5% | 2126.28 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP241610MG
Encompass_Preferred
Gastrin I, rat (Rat Gastrin-17) is a peptide hormone that potently stimulates gastric acid secretion. This product is for research use only.
  • Potent stimulator of gastric acid secretion
  • Appears as a white to off-white solid
  • Requires sealed storage, away from moisture and light, under nitrogen
  • Stable as powder at -80°C for 2 years or -20°C for 1 year
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-24239