Learn More
CPC Scientific H-Ala-Pro-Val-Ser-Val-Gly-Gly-Gly-Thr-Val-Leu-Ala-Lys-Met-Tyr-Pro-Arg-Gly-Asn-His-Trp-Ala-Val-Gly-His-Leu-Met-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific GAST004A
Encompass
ONE-LETTER SEQUENCE: APVSVGGGTVLAKMYPRGNHWAVGHLM-NH2MOLECULAR FORMULA: C126H198N38O31S2MOLECULAR WEIGHT:2805.3STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [74815-57-9], RESEARCH AREA: GastrointestinalREFERENCES: T. J. McDonald et al., Biochem. Biophys. Res. Commun., 90, 227 (1979)
Catalog No. 50-210-9321