Learn More
TARGETMOL CHEMICALS INC GAT107 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T274045MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. GAT107 is a novel and potent (alpha)7 nicotinic cholinergic receptor modifier agonist and modulator with anti-inflammatory activity that reverses injury perception in a mouse model of neuropathic pain. purity: 98%
Catalog No. 50-258-0617